Order Entry
Export
Orders LinkContactUsLinkComponent
CDC53-Py2-HA 1 ml 1 * 1 mL

CDC53-Py2-HA 1 ml 1 * 1 mL

Catalog # ORBGORB-BVC-10243
Supplier:   ORBIGEN
CDC53-Py2-HA 1 ml 1 * 1 mL
Catalog # ORBGORB-BVC-10243
Supplier:  ORBIGEN
CDC53-Py2-HA 1 ml 1 * 1 mL
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents