Order Entry
Export
Orders LinkContactUsLinkComponent
His6-MCM6 1 ml 1 * 1 mL

His6-MCM6 1 ml 1 * 1 mL

Catalog # ORBGORB-BVC-10162
Supplier:   ORBIGEN
His6-MCM6 1 ml 1 * 1 mL
Catalog # ORBGORB-BVC-10162
Supplier:  ORBIGEN
His6-MCM6 1 ml 1 * 1 mL
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents