Order Entry
Export
Orders LinkContactUsLinkComponent
HPV33-E6 oncogene 1 ml 1 * 1 mL

HPV33-E6 oncogene 1 ml 1 * 1 mL

  ORBGORB-BVC-10123
 :   ORBIGEN
HPV33-E6 oncogene 1 ml 1 * 1 mL
  ORBGORB-BVC-10123
 :  ORBIGEN
HPV33-E6 oncogene 1 ml 1 * 1 mL
 
 
   
1 * 1 mL