Order Entry
Export
Orders LinkContactUsLinkComponent
replication factor C p140 (RFCp140) 1 1 * 1 mL

replication factor C p140 (RFCp140) 1 1 * 1 mL

Catalog # ORBGORB-BVC-10094
Supplier:   ORBIGEN
replication factor C p140 (RFCp140) 1 1 * 1 mL
Catalog # ORBGORB-BVC-10094
Supplier:  ORBIGEN
replication factor C p140 (RFCp140) 1 1 * 1 mL
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents