Order Entry
Export
Orders LinkContactUsLinkComponent
alpha - p53 (Do-7) 0,1 ml 1 * 0,1 mL

alpha - p53 (Do-7) 0,1 ml 1 * 0,1 mL

Catalog # ORBGBCR-CM042A
Supplier:   ORBIGEN
alpha - p53 (Do-7) 0,1 ml 1 * 0,1 mL
Catalog # ORBGBCR-CM042A
Supplier:  ORBIGEN
alpha - p53 (Do-7) 0,1 ml 1 * 0,1 mL
Price
Inquire For Price
Log in for availability
1 * 0,1 mL
Product Details & Documents