Order Entry
Export
Orders LinkContactUsLinkComponent
SRK® Surfaces and Equipment Microbial Testing Devices

SRK® Surfaces and Equipment Microbial Testing Devices

SRK® - tube with 10 ml of SRK® solution + 1 separate large plastic applicator, viscose tip; Rayon wrapped regular tip; 25×10, individually wrapped; 16×100 skirted sferic bottom tube, 10 ml, Green
Catalog # 710-0474
Supplier:   Copan
SRK® Surfaces and Equipment Microbial Testing Devices
Catalog # 710-0474
Supplier:  Copan
SRK® Surfaces and Equipment Microbial Testing Devices
SRK® Surfaces and Equipment Microbial Testing Devices

SRK® - tube with 10 ml of SRK® solution + 1 separate large plastic applicator, viscose tip; Rayon wrapped regular tip; 25×10, individually wrapped; 16×100 skirted sferic bottom tube, 10 ml, Green

Price
29,20 €
Log in for availability
1 * 25 items
Product Details & Documents

Our Swab Rinse Kits line includes comprehensive tailor-made collection and devices for the detection of environmental microbial contamination in a broad range of environments, such as food production, pharmaceutical sites, and hospital wards. Inspectors and Quality Assurance personnel can use SRK® products for routine surface monitoring within manufacturing areas, hospital surfaces, and equipment.

  • Optimal recovery rate: Choose the sampling probe that best suits your needs: regular FLOQSwabs® and CLASSIQSwabs™, plastic/viscose applicator, or sponge spatula

    Compliance: SRK® kits are designed compliant with ISO 18593:2018, ISO 18593:2004, ISO 17604:2003, and EU directive 2001/471/E.C

    Media choice for enhanced preservation: Choose between an isotonic solution or a wide range of selective and enrichment broths: Half Fraiser, RVS, BPW 1%, TSB, UNS, and Letheen broth

    Sterile sampling frames: the definition of a set sampling area is essential for microbial bioburden validations

    Sampling frames will make your surface sampling match that standard

This product line groups a variety of product codes, each optimised to address specific needs.

Specifications
  • Description:
    SRK® - tube with 10 ml of SRK® solution + 1 separate large plastic applicator, viscose tip; Rayon wrapped regular tip; 25×10, individually wrapped; 16×100 skirted sferic bottom tube, 10 ml
  • Seq:
    5
  • Cap colour:
    Green