Decappers, E-Z Decappers™, WHEATON®
Decapper for 11 mm crimp caps
Catalog # WHEAW225351
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
These decappers are used to remove aluminium seals from bottles and vials with crimp or serum finish.
- Cushioned ergonomic handle reduces hand fatigue
- Labeled for quick size identification
- Autoclavable
Specifications
- Description:Decapper for 11 mm crimp caps
- Seq:02