Orders LinkContactUsLinkComponent
Decappers, E-Z Decappers™, WHEATON®
Decappers, E-Z Decappers™, WHEATON®
Decapper for 8 mm crimp caps
Catalog # WHEAW225350
Supplier:   DWK Life Sciences
Decappers, E-Z Decappers™, WHEATON®
Catalog # WHEAW225350
Supplier:  DWK Life Sciences
Decappers, E-Z Decappers™, WHEATON®
Decappers, E-Z Decappers™, WHEATON®
Decapper for 8 mm crimp caps
Price
1 170,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These decappers are used to remove aluminium seals from bottles and vials with crimp or serum finish.
  • Cushioned ergonomic handle reduces hand fatigue
  • Labeled for quick size identification
  • Autoclavable
Specifications
  • Description:
    Decapper for 8 mm crimp caps
  • Seq:
    01