Order Entry
Export
Orders LinkContactUsLinkComponent
pH buffers, DuraCal

pH buffers, DuraCal

10,01, ±0,02, Carbonate, Yellow, Plastic bottle, 250 mL
Catalog # HAMI238321
Supplier:   HAMILTON BONADUZ
pH buffers, DuraCal
Catalog # HAMI238321
Supplier:  HAMILTON BONADUZ
CAS Number:  
pH buffers, DuraCal
pH buffers, DuraCal

10,01, ±0,02, Carbonate, Yellow, Plastic bottle, 250 mL

CAS Number:  
Price
24,90 €
Log in for availability
1 * 250 mL
Product Details & Documents

For full information concerning health and safety data please contact Avantor or Hamilton directly.

  • Label includes expiration date, batch number and actual value
  • Traceable to NIST and PTB, certificates available to download
  • Stable solutions
  • Certified pH value from a DAkkS laboratory accredited for pH measurement
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Carbonate
  • Pk:
    250 mL
  • Colour:
    Yellow
  • pH accuracy:
    ±0,02
  • pH value:
    10,01

Specifications