VWR®, Beaker Cups, Disposable
150 ml, 65 mm
611-1367
PS.
- Suitable for use in various sample dispensers
- Beakers with capacity of 5 and 20 ml are recommended for use in flame photometric analyses
Economical, multipurpose beaker for a very wide range of uses.
- Capacity:150 ml
- Height:65 mm
- Seq:06



