Microscope slides, UniMark®
Boxes (50) of 50 slides in an aluminium bag, Green
Catalog # 630-1600
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Microslide green 90 ground edge alu unimark made of soda lime glass of 3hydrolytic class in compliance with din iso 8037/1,dimensions: approx. 76 x 26 mm, thickness approx. 1 mm. 1 * 2.500 items Catalog #MARI0704207
Product MARI0704207 is a replacement for 630-1600 but varies by Brand, Packaging or Quantity.
Product Details & Documents
The bright printed colours of UniMark® slides frosted ends contrast well with the markings.
- Frosted end for labelling
- Soda-lime glass, Hydrolytic Class 3
- Different colours to identify preparations securely
- Thin coating to facilitate the processing of slides in automated systems
W×D×H: 76×26×1 mm
For in vitro diagnostic (IVD) applications.
Documents
Certificates
Search by Lot NumberCoC_MARI1000612_823410
Conformity declaration
MARI0704102_TDS_EN
Datasheet
Specifications
- Colour:Green
- Description:Boxes (50) of 50 slides in an aluminium bag
- Seq:09