Orders LinkContactUsLinkComponent
Eco caps
Eco caps
Replacement male Luer plug, PP
Catalog # JOURJR-070350-10
Supplier:   VICI JOUR RESEARCH
Eco caps
Catalog # JOURJR-070350-10
Supplier:  VICI JOUR RESEARCH
Eco caps
Replacement male Luer plug, PP
Price
30,80 €
Log in for availability
1 * 10 items
Product Details & Documents
The Eco Cap consists of a three-hole ETFE insert with a polyethylene collar.
  • Compatible with all standard GL 45 thread bottles
  • Also available as bottle kit
There is no threading for fittings on the inside of the tubing ports, the tubing can be easily inserted.
Delivery information: The Eco Cap is supplied with one cap, polypropylene male Luer plug. The kit also includes safety-coated glass bottle with a option of 1 or 2 L.
Specifications
  • Description:
    Replacement male Luer plug, PP
  • Seq:
    04