ContactUsLinkComponent
Thermometer precision temperature range -5 to 200:0,2C enclosed scale total immersion length 550mm mercury filled with DKD certifi c ate Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be 1 * 1 items
Catalog # AMARL25954DKD4
Supplier:  Amarell
Thermometer precision temperature range -5 to 200:0,2C enclosed scale total immersion length 550mm mercury filled with DKD certifi c ate Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be 1 * 1 items
Catalog # AMARL25954DKD4
Supplier:  Amarell