Sterile containers, Falcon®
220 ml, without lid, 20/bag, 500/Case
Supplier: Corning
Jump to:
Product Details & Documents
PP, sterile, with or without PE lid.
- Moulded-in graduations for easy measurement
- Inert and chemically resistant to commonly used laboratory reagents at room temperature
Convenient, single-use containers for collection, transport and storage of a wide variety of specimens.
Polyethylene lid is frosted for easy marking, and fits both container sizes.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Packed:20/bag, 500/Case
- Capacity:220 ml
- Description:without lid
- Seq:03
- Cat. No.:392-0396
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Low Manufacturing Impact