Orders LinkContactUsLinkComponent
Disposable earplugs, Max®
Disposable earplugs, Max®
Earplugs, with cord, Red, U*
Catalog # 111-0139
Supplier:   HONEYWELL SAFETY
Disposable earplugs, Max®
Catalog # 111-0139
Supplier:  HONEYWELL SAFETY
Disposable earplugs, Max®
Disposable earplugs, Max®
Earplugs, with cord, Red, U*
Price
63,00 €
Log in for availability
1 * 100 PAIR
Product Details & Documents
Max® earplugs offer superior noise blocking performance in a disposable earplug.
  • Bell shape provides maximum ear comfort
  • Improved design is easier to insert
  • Smooth, soil resistant covering prevents dirt penetrating the surface prior to insertion
SNR: 37 dB
SNR: 37 dB
EN 352-2:2002
Certifications: EN 352-2:2002
Specifications
  • Colour:
    Red
  • Description:
    Earplugs, with cord
  • Seq:
    02
  • Size:
    U*