Order Entry
Export
Orders LinkContactUsLinkComponent
pH buffers, DuraCal

pH buffers, DuraCal

5,00, ±0,02, Colourless, Plastic bottle, 500 mL
  HAMI238275
pH buffers, DuraCal
  HAMI238275
 :  
pH buffers, DuraCal
pH buffers, DuraCal

5,00, ±0,02, Colourless, Plastic bottle, 500 mL

 :  
 
34,40 €
   
1 * 500 mL
 

For full information concerning health and safety data please contact Avantor or Hamilton directly.

  • Label includes expiration date, batch number and actual value
  • Traceable to NIST and PTB, certificates available to download
  • Stable solutions
  • Certified pH value from a DAkkS laboratory accredited for pH measurement

Documents

 

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Colour:
    Colourless
  • pH accuracy:
    ±0,02
  • pH value:
    5,00