Order Entry
Export
Orders LinkContactUsLinkComponent
 

Pasteur pipettes, disposable, LDPE

3,3 ml, 155 mm, - ml
  SEMA2291
 :   SEMADENI
Pasteur pipettes, disposable, LDPE
  SEMA2291
 :  SEMADENI
Pasteur pipettes, disposable, LDPE
Pasteur pipettes, disposable, LDPE

3,3 ml, 155 mm, - ml

 
83,80 €
   
1 * 500 items
 

LDPE, disposable, non-sterile.

  • Almost unbreakable transfer pipettes
  • Good chemical resistance and can be used at low temperatures
  • Pipettes are made from one piece: No risk of leakage
  • Dustproof packaged in convenient standard boxes
 
  • Length:
    155 mm
  • Seq:
    01
  • Graduation:
    - ml
  • Bulb draw:
    3,3 ml