Orders LinkContactUsLinkComponent
Distilling bridges
Distilling bridges
200 mm, 19/26 NS
Catalog # WITG1.205.260
Supplier:   witeg Labortechnik
Distilling bridges
Catalog # WITG1.205.260
Supplier:  witeg Labortechnik
Distilling bridges
Distilling bridges
200 mm, 19/26 NS
Price
108,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two vertical NS cones and splash head.
Specifications
  • Cone size:
    19/26 NS
  • Jacket length:
    200 mm