3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate
100 mL
Catalog # ROCKTMBM-100
Supplier: Rockland Immunochemicals
3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate
Catalog # ROCKTMBM-100
Supplier: Rockland Immunochemicals
CAS Number: 54827-17-7
Jump to:
Product Details & Documents
In the presence of peroxidases, TMB substrates can act as an electron donor for the conversion of peroxides to water.
- Reaction can be halted with acid to change the TMB to yellow
- Soluble blue end product read at 370 nm or 655 nm
- Incubation time varies depending on the assay conditions
TMB ELISA peroxidase substrate (3, 3', 5, 5'-tetramethylbenzidine) is a chromogenic substrate used for antibody reactivity in ELISA experiments.
TMB ELISA peroxidase substrate is designed for use in immunology and cancer research.
Delivery information: Supplied as a ready to use clear to very light blue colour liquid.
Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
- CAS Number:54827-17-7
- Formula:C₁₆H₂₀N₂
- MDL Number:MFCD00007748
- Molecular Weight:240.35 g/mol
Documents
TMBE-100_.pdf
Datasheet
Specifications
Specifications
- Pk:100 mL



