Order Entry
Export
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate

3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate

100 mL
Catalog # ROCKTMBE-100
3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate
Catalog # ROCKTMBE-100
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate
3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate

100 mL

CAS Number:  54827-17-7
Price
239,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

In the presence of peroxidases, TMB substrates can act as an electron donor for the conversion of peroxides to water.

  • Reaction can be halted with acid to change the TMB to yellow
  • Soluble blue end product read at 370 nm or 655 nm
  • Incubation time varies depending on the assay conditions

TMB ELISA peroxidase substrate (3, 3', 5, 5'-tetramethylbenzidine) is a chromogenic substrate used for antibody reactivity in ELISA experiments.

TMB ELISA peroxidase substrate is designed for use in immunology and cancer research.

Delivery information: Supplied as a ready to use clear to very light blue colour liquid.

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol

Documents

Specifications

Specifications

  • Pk:
    100 mL

Specifications