NPP ELISA alkaline phosphatase substrate
1X Working Buffer for NPP-10, 500 mL
Catalog # ROCKNPP-B500
Supplier: Rockland Immunochemicals
NPP ELISA alkaline phosphatase substrate
Catalog # ROCKNPP-B500
Supplier: Rockland Immunochemicals
CAS Number:
Jump to:
Product Details & Documents
NPP ELISA alcaline phosphatase substrate is a chromogenic substrate used in immunosorbent assays.
Phosphatase-conjugated detection antibodies can act upon the pNPP to convert it to a yellow para-nitrophenol. This yellow product can be measured on a spectrophotometer at 405 nm.
Ordering information: Diluent buffer NPP-B500 for NPP-10 has to be ordered separately.
Caution: Both the substrate and buffer have to be stored at 4 °C prior to opening and should be protected from moisture and light.
Documents
NPP-B500_
Datasheet
Specifications
Specifications
- Pk:500 mL
- Description:1X Working Buffer for NPP-10



