Order Entry
Export
Orders LinkContactUsLinkComponent
 

Conjugate stabilisers

ALP Conjugate Stabiliser, 1.000 mL
Catalog # ROCKMB-061-1000
Conjugate stabilisers
Catalog # ROCKMB-061-1000
CAS Number:  
Conjugate stabilisers

ALP Conjugate Stabiliser, 1.000 mL

CAS Number:  
Price
447,00 €
Log in for availability
1 * 1.000 mL
Product Details & Documents

Conjugate stabilisers are used with horseradish peroxidase (HRP) and calf intestinal alkaline phosphatase conjugated primary or secondary antibodies or other proteins or compounds in solution. The components of the stabiliser (proprietary) are unique and stabilise both the antibody and enzyme portions of the conjugate.

This conjugate stabiliser prevents the loss of catalytic activity and maintains the active conformation of the antibody portion of the conjugate. This 1X diluent buffer contains buffered saline along with proprietary reagents to stabilise and preserve both HRP and ALP conjugated primary or secondary antibodies in solution. A proprietary combination of preservatives is used that is azide- and mercury-free.

 : Supplied as ready to use sterile filtered liquid.

Specifications

Specifications

  • Pk:
    1.000 mL
  • Description:
    ALP Conjugate Stabiliser

Specifications