Order Entry
Export
Orders LinkContactUsLinkComponent
Hydroxyectoine

Hydroxyectoine

Glass bottle for solids, 100 g
Catalog # BMOL55460-4
Hydroxyectoine
Catalog # BMOL55460-4
CAS Number:  165542-15-4
Hydroxyectoine

Glass bottle for solids, 100 g

CAS Number:  165542-15-4
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:165542-15-4
  • Formula:C₆H₁₀N₂O₃
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications