n-Dodecyl-β-D-maltoside
Glass bottle for solids, 5 g
Catalog # BMOL56366-2
Supplier: BIOMOL RESEARCH LABORATORIES
n-Dodecyl-β-D-maltoside
Catalog # BMOL56366-2
Supplier: BIOMOL RESEARCH LABORATORIES
CAS Number: 69227-93-6
Jump to:
Product Details & Documents
Synonyms: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside
- CAS Number:69227-93-6
- Formula:C₂₄H₄₆O₁₁
- MDL Number:MFCD00043012
- Molecular Weight:510.6 g/mol
- Storage Temperature:Freezer
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



