Order Entry
Export
Orders LinkContactUsLinkComponent
Glycerine

Glycerine

Glass bottle, 1 L
Catalog # BMOL50405-1
Glycerine
Catalog # BMOL50405-1
CAS Number:  56-81-5
Glycerine
Glycerine

Glass bottle, 1 L

CAS Number:  56-81-5
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
  • CAS Number:56-81-5
  • Formula:HOCH₂CH(OH)CH₂OH
  • MDL Number:MFCD00004722
  • Molecular Weight:92.09 g/mol
  • Merck Index:13,04497
  • Boiling Pt:290 °C (1013 hPa)
  • Melting Pt:18.6 °C
  • Density:1.26 g/cm³ (20 °C)
  • Flash Pt:177 °C
  • Storage Temperature:Ambient
  • EINECS:200-289-5
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications