Glycerine
Glass bottle, 5 L
Catalog # BMOL50405-2
Supplier: BIOMOL RESEARCH LABORATORIES
Jump to:
Product Details & Documents
Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
- CAS Number:56-81-5
- Formula:HOCH₂CH(OH)CH₂OH
- MDL Number:MFCD00004722
- Molecular Weight:92.09 g/mol
- Merck Index:13,04497
- Boiling Pt:290 °C (1013 hPa)
- Melting Pt:18.6 °C
- Density:1.26 g/cm³ (20 °C)
- Flash Pt:177 °C
- Storage Temperature:Ambient
- EINECS:200-289-5
Specifications
Specifications
- Pack type:Glass bottle
- Pk:5 L



