Order Entry
Export
Orders LinkContactUsLinkComponent
Ethynyltributyltin 96%

Ethynyltributyltin 96%

Glass bottle for solids, 1 g
Catalog # L20178.03
Toxic
Health
Irritant
Ethynyltributyltin 96%
Catalog # L20178.03
CAS Number:  994-89-8
Ethynyltributyltin 96%
Ethynyltributyltin 96%

Glass bottle for solids, 1 g

CAS Number:  994-89-8
Price
72,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 96% CAS No.: 994-89-8 Molecular Formula: C14H28Sn Formula Weight: 315.09 Storage Temperature: 2°C to 8°C Flash Point: 73°(163°F) Refractive Index: 1.4770 Boiling Point: 70°/0.2mm Beilstein Reference: 3537659 UN No.: 2788 MDL No.: MFCD00009420

Synonyms:  Ethynyltri-n-butylstannane, Tributylethynylstannane
  • CAS Number:994-89-8
  • Formula:C₁₄H₂₈Sn
  • MDL Number:MFCD00009420
  • Molecular Weight:315.09 g/mol
  • Boiling Pt:70 °C (0.2 mmHg)
  • Density:1.080
  • Flash Pt:73 °C (163 °F)
  • UN:2788
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications