2,3-Dimethoxypyridine
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 52605-97-7 Molecular Formula: C7H9NO2 Formula Weight: 139.15 Refractive Index: 1.5205 Boiling Point: 98-100°/16mm Beilstein Reference: 1526067
- CAS Number:52605-97-7
- Formula:C₇H₉NO₂
- MDL Number:MFCD03095043
- Molecular Weight:139.15 g/mol
- Boiling Pt:98…100 °C (16 mmHg)
- Density:1.104
- Storage Temperature:Ambient
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



