Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,3-Dibromopyridine ≥97%

Glass bottle for solids, 1 g
Catalog # L19527.03
Toxic
Irritant
2,3-Dibromopyridine ≥97%
Catalog # L19527.03
CAS Number:  13534-89-9
2,3-Dibromopyridine ≥97%
2,3-Dibromopyridine ≥97%

Glass bottle for solids, 1 g

CAS Number:  13534-89-9
Price
43,30 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 97% CAS No.: 13534-89-9 Molecular Formula: C5H3Br2N Formula Weight: 236.90 Melting Point: 58-60° Boiling Point: 249-250° MDL No.: MFCD00234014

  • CAS Number:13534-89-9
  • Formula:C₅H₃Br₂N
  • MDL Number:MFCD00234014
  • Molecular Weight:236.89 g/mol
  • Boiling Pt:249…250 °C
  • Melting Pt:58…60 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications