Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cdk-cyclin a reaction buffer - 5.0 ml 1 * 5 ml
Catalog # NEWEB6025S
Supplier:
NEW ENGLAND BIOLABS
Expand image
Cdk-cyclin a reaction buffer - 5.0 ml 1 * 5 ml
Catalog # NEWEB6025S
Supplier:
NEW ENGLAND BIOLABS