Dichloromethane ≥99% stabilised
Glass bottle, 1.000 mL
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99% stabilised
Catalog # L13089.AU
Supplier: Thermo Fisher Scientific
CAS Number: 75-09-2
Jump to:
Product Details & Documents
Stabilized with ca. 50 ppm amylene
Purity: 99+% CAS No.: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93 Refractive Index: 1.4244 Odour: Sweet Boiling Point: 39-40° Beilstein Reference: 1730800 Merck Reference: 14,6063 UN No.: 1593 MDL No.: MFCD00000881
Synonyms: Methylene chloride, DCM
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:14,6063
- Boiling Pt:39…40 °C
- Melting Pt:–95 °C
- Density:1.325
- Storage Temperature:Ambient
- EINECS:200-838-9
- ADR:6.1,III
- :Stabilised with 2-methyl-2-butene (iso-amylene) 50 ppm
- UN:1593
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1.000 mL



