Order Entry
Export
Orders LinkContactUsLinkComponent
1,2-Ethanedithiol ≥98%

1,2-Ethanedithiol ≥98%

Glass bottle for solids, 500 g
Catalog # L12865.36
Toxic
Flammable
Irritant
1,2-Ethanedithiol ≥98%
Catalog # L12865.36
CAS Number:  540-63-6
1,2-Ethanedithiol ≥98%
1,2-Ethanedithiol ≥98%

Glass bottle for solids, 500 g

CAS Number:  540-63-6
Price
278,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98+% CAS No.: 540-63-6 Molecular Formula: C2H6S2 Formula Weight: 94.20 Flash Point: 45°(113°F) Refractive Index: 1.5580 Odour: Stench Melting Point: -41° Boiling Point: 144-146° Beilstein Reference: 505946 Merck Reference: 14,3725 UN No.: 3071 MDL No.: MFCD00004892

Synonyms:  1.2-Ethanedithiol, 1,2-Dimercaptoethane
  • CAS Number:540-63-6
  • Formula:C₂H₆S₂
  • MDL Number:MFCD00004892
  • Molecular Weight:94.2 g/mol
  • Merck Index:14,3725
  • Boiling Pt:144…146 °C
  • Melting Pt:–41 °C
  • Density:1.123
  • Flash Pt:45 °C (113 °F)
  • Storage Temperature:Ambient
  • EINECS:208-752-3
  • UN:3071
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications