Dissolution vessels, QLA
QLA clear glass dissolution vessel without ring, 1000 ml
Catalog # QLAAGLA900-DK
Supplier: QUALITY LAB ACCESSORIES
Jump to:
Product Details & Documents
Dissolution vessels are extremely important because the vessels associated with various dissolution baths are not dimensionally the same. Replacing a vessel with one not specifically designed for the instrument could impair vessel centering.
- Made of high quality borosilicate glass or plastic
- Dissolution vessels comply with both USP (711) and dimensional specifications
Also larger or smaller vessel inside diameters can change hydrodynamics. Both variables could impact dissolution rates.
Specifications
- Description:QLA clear glass dissolution vessel without ring, 1000 ml
- Seq:09