Orders LinkContactUsLinkComponent
Dissolution vessels, QLA
Dissolution vessels, QLA
QLA clear glass dissolution vessel, serialised, 1000 ml
Catalog # QLAAGLA900-01
Dissolution vessels, QLA
Catalog # QLAAGLA900-01
Dissolution vessels, QLA
Dissolution vessels, QLA
QLA clear glass dissolution vessel, serialised, 1000 ml
Price
298,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Dissolution vessels are extremely important because the vessels associated with various dissolution baths are not dimensionally the same. Replacing a vessel with one not specifically designed for the instrument could impair vessel centering.
  • Made of high quality borosilicate glass or plastic
  • Dissolution vessels comply with both USP (711) and dimensional specifications
Also larger or smaller vessel inside diameters can change hydrodynamics. Both variables could impact dissolution rates.
Specifications
  • Description:
    QLA clear glass dissolution vessel, serialised, 1000 ml
  • Seq:
    04