Toluene ≥99%
Glass bottle, 1.000 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 108-88-3 Molecular Formula: C7H8 Formula Weight: 92.14 Flash Point: 4°(40°F) Refractive Index: 1.4967 Odour: Aromatic Melting Point: ca -93° Boiling Point: 111° Beilstein Reference: 635760 Merck Reference: 14,9529 UN No.: 1294 MDL No.: MFCD00008512
Synonyms: Methylbenzene, Phenylmethane, FM approval
- CAS Number:108-88-3
- Formula:C₆H₅CH₃
- MDL Number:MFCD00008512
- Molecular Weight:92.14 g/mol
- Merck Index:14,9529
- Boiling Pt:111 °C
- Melting Pt:∼–93 °C
- Density:0.867
- Flash Pt:4 °C (40 °F)
- Storage Temperature:Ambient
- EINECS:203-625-9
- UN:1294
- ADR:3,II
- REACH:05-2114615130-69
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1.000 mL



