Ethyl acetate ≥99%
Glass bottle, 1.000 mL
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99%
Catalog # L10925.AU
Supplier: Thermo Fisher Scientific
CAS Number: 141-78-6
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 141-78-6 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: -3°(26°F) Refractive Index: 1.3720 Odour: Sweet Melting Point: -84° Boiling Point: 76-77° Beilstein Reference: 506104 Merck Reference: 14,3757 UN No.: 1173 MDL No.: MFCD00009171
Synonyms: Acetic acid ethyl ester
- CAS Number:141-78-6
- Formula:CH₃COOC₂H₅
- MDL Number:MFCD00009171
- Molecular Weight:88.11 g/mol
- Merck Index:14,3757
- Boiling Pt:76…77 °C
- Melting Pt:–84 °C
- Density:0.902
- Flash Pt:–3 °C (26 °F)
- Storage Temperature:Ambient
- EINECS:205-500-4
- UN:1173
- ADR:3,II
- REACH:01-2119475103-46
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1.000 mL
- Cat. No.:L10925.AU
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health



