Linoleic acid 95%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 95% CAS No.: 60-33-3 Molecular Formula: C18H32O2 Formula Weight: 280.45 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4699 Odour: Weak, characteristic Melting Point: -12° Boiling Point: 230°/16mm Beilstein Reference: 1727101 Merck Reference: 14,5505 MDL No.: MFCD00064241
Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
- CAS Number:60-33-3
- Formula:C₁₈H₃₂O₂
- MDL Number:MFCD00064241
- Molecular Weight:280.45 g/mol
- Merck Index:14,5505
- Boiling Pt:230 °C (16 mmHg)
- Melting Pt:–12 °C
- Density:0.902
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Refrigerator
- EINECS:200-470-9
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



