Order Entry
Export
Orders LinkContactUsLinkComponent
2-Nitro-m-xylene ≥99%

2-Nitro-m-xylene ≥99%

Glass bottle for solids, 500 g
Catalog # L05546.36
Irritant
2-Nitro-m-xylene ≥99%
Catalog # L05546.36
CAS Number:  81-20-9
2-Nitro-m-xylene ≥99%
2-Nitro-m-xylene ≥99%

Glass bottle for solids, 500 g

CAS Number:  81-20-9
Price
136,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 81-20-9 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Flash Point: 87°(188°F) Refractive Index: 1.5220 Melting Point: 14-16° Boiling Point: 225-227° Beilstein Reference: 2046066 UN No.: 1665 MDL No.: MFCD00007181

Synonyms:  2-Nitro-m-xylene, 1,3-Dimethyl-2-nitrobenzene, 2,6-Dimethylnitrobenzene
  • CAS Number:81-20-9
  • Formula:C₈H₉NO₂
  • MDL Number:MFCD00007181
  • Molecular Weight:151.16 g/mol
  • Boiling Pt:225…227 °C
  • Melting Pt:14…16 °C
  • Density:1.110
  • Flash Pt:87 °C (188 °F)
  • Storage Temperature:Ambient
  • EINECS:201-333-6
  • UN:1665
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications