Accessories for PURELAB Flex Systems
Filter 0,2 µ: removes particulates, colloids and bacteria for analytical grade water applications.
For PURELAB® flex 2 and 3.
For PURELAB® flex 2 and 3.
Catalog # 171-0476
Supplier: ELGA LabWater
Jump to:
Product Details & Documents
Specifications
- Description:Filter 0,2 µ: removes particulates, colloids and bacteria for analytical grade water applications.
For PURELAB® flex 2 and 3.