Orders LinkContactUsLinkComponent
Dimethylphenylsilane ≥97%
Glass bottle for solids, 50 g
  L04558.18
Dimethylphenylsilane ≥97%
  L04558.18
 :  766-77-8
Dimethylphenylsilane ≥97%
Dimethylphenylsilane ≥97%
Glass bottle for solids, 50 g
 :  766-77-8
182,00 €
1 * 50 g
Purity: 97% CAS No.: 766-77-8 Molecular Formula: C8H12Si Formula Weight: 136.27 Flash Point: 35°(95°F) Refractive Index: 1.4970 Melting Point: -124° Boiling Point: 156-157° Beilstein Reference: 2204906 UN No.: 1993 MDL No.: MFCD00008257
 :  Dimethyl(phenyl)silane
  •  :766-77-8
  •  :C₈H₁₂Si
  •  :MFCD00008257
  •  :136.27 g/mol
  •  :156…157 °C
  •  :–124 °C
  •  :0.879
  •  :35 °C (95 °F)
  •  :Ambient
  •  :212-170-5
  •  :3,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g