Order Entry
Export
Orders LinkContactUsLinkComponent
Dimethylphenylsilane ≥97%

Dimethylphenylsilane ≥97%

Glass bottle for solids, 10 g
Catalog # L04558.09
Flammable
Dimethylphenylsilane ≥97%
Catalog # L04558.09
CAS Number:  766-77-8
Dimethylphenylsilane ≥97%
Dimethylphenylsilane ≥97%

Glass bottle for solids, 10 g

CAS Number:  766-77-8
Price
56,30 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 97% CAS No.: 766-77-8 Molecular Formula: C8H12Si Formula Weight: 136.27 Flash Point: 35°(95°F) Refractive Index: 1.4970 Melting Point: -124° Boiling Point: 156-157° Beilstein Reference: 2204906 UN No.: 1993 MDL No.: MFCD00008257

Synonyms:  Dimethyl(phenyl)silane
  • CAS Number:766-77-8
  • Formula:C₈H₁₂Si
  • MDL Number:MFCD00008257
  • Molecular Weight:136.27 g/mol
  • Boiling Pt:156…157 °C
  • Melting Pt:–124 °C
  • Density:0.879
  • Flash Pt:35 °C (95 °F)
  • Storage Temperature:Ambient
  • EINECS:212-170-5
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications