Order Entry
Export
Orders LinkContactUsLinkComponent
 

Fibronectin-coated flasks, Corning® BioCoat®

Cell culture flask, 750 ml, 175 cm², Plug seal screw cap, Straight
Catalog # 734-0184
   Low Manufacturing Impact
Supplier:   Corning
Fibronectin-coated flasks, Corning® BioCoat®
Catalog # 734-0184
Supplier:  Corning
   Low Manufacturing Impact
Fibronectin-coated flasks, Corning® BioCoat®
Fibronectin-coated flasks, Corning® BioCoat®

Cell culture flask, 750 ml, 175 cm², Plug seal screw cap, Straight

Price
477,58 €
Log in for availability
1 * 5 items
Product Details & Documents

Human Fibronectin (HFN) is a widely distributed glycoprotein that is used as a substrate to promote attachment of cells through its central-binding domain RGD sequence. HFN is a product of most mesenchymal and epithelial cells and is present in both the ECM and plasma. The principal function of HFN appears to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis

Applications include promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assays, studies of the effects of fibronectin on cell behaviour, and improving survival of primary cells in culture.

Documents

Specifications
  • Closure type:
    Plug seal screw cap
  • Growth area:
    175 cm²
  • Capacity:
    750 ml
  • Description:
    Cell culture flask
  • Seq:
    02
  • Neck:
    Straight
  • Cat. No.:
    734-0184
  • Environmental certification:
    ISO 14001 - Bedford
  • Environmentally Preferable:
    Low Manufacturing Impact