7-Hydroxycoumarin ≥98%
100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 93-35-6 Molecular Formula: C9H6O3 Formula Weight: 162.14 Melting Point: 228-232° Beilstein Reference: 127683 Merck Reference: 14,9847 MDL No.: MFCD00006878
Synonyms: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
- CAS Number:93-35-6
- Formula:C₉H₆O₃
- MDL Number:MFCD00006878
- Molecular Weight:162.14 g/mol
- Merck Index:14,9847
- Boiling Pt:382 °C (1013 hPa)
- Melting Pt:228…232 °C
- Storage Temperature:Ambient
- EINECS:202-240-3
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pk:100 g



