Order Entry
Export
Orders LinkContactUsLinkComponent
 

7-Hydroxycoumarin ≥98%

25 g
Catalog # L04082.14
Irritant
7-Hydroxycoumarin ≥98%
Catalog # L04082.14
CAS Number:  93-35-6
7-Hydroxycoumarin ≥98%

25 g

CAS Number:  93-35-6
Price
58,80 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 93-35-6 Molecular Formula: C9H6O3 Formula Weight: 162.14 Melting Point: 228-232° Beilstein Reference: 127683 Merck Reference: 14,9847 MDL No.: MFCD00006878

Synonyms:  Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
  • CAS Number:93-35-6
  • Formula:C₉H₆O₃
  • MDL Number:MFCD00006878
  • Molecular Weight:162.14 g/mol
  • Merck Index:14,9847
  • Boiling Pt:382 °C (1013 hPa)
  • Melting Pt:228…232 °C
  • Storage Temperature:Ambient
  • EINECS:202-240-3
Specifications

Specifications

  • Pk:
    25 g

Specifications