Test Kits, BOD, Spectroquant®, Supelco®
BOD Spectroquant® standard **, 210 mg/l, 10 Tests
Supplier: Merck
Jump to:
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*3 BOD nutrient salt mixture 1.00688 required
Documents
Specifications
Specifications
- Concentration:210 mg/l
- Pk:10 Tests
- Description:BOD Spectroquant® standard **
- Cat. No.:1.00718.0001
- M-Clarity quality level:MQ100
Specifications
Accessories
Product Information
- Catalog #TypeAvailabilityPrice
- Oxygen reaction tube for 1.14662.0001 (MTC)Oxygen reaction tube for 1.14662.0001 (MTC)1.14663.0001MTCLog in for availability1 * 1 items - 97,40 €