Orders LinkContactUsLinkComponent
Test Kits, BOD, Spectroquant®, Supelco®
Test Kits, BOD, Spectroquant®, Supelco®
BOD cell test*³, 0,5 - 3000 mg/l, Winkler (modified), Prove, Nova 30/60, Multy, Move, 50 Tests
Catalog # 1.00687.0001
Supplier:   Merck
Test Kits, BOD, Spectroquant®, Supelco®
Catalog # 1.00687.0001
Supplier:  Merck
CAS Number:  
Test Kits, BOD, Spectroquant®, Supelco®
Test Kits, BOD, Spectroquant®, Supelco®
BOD cell test*³, 0,5 - 3000 mg/l, Winkler (modified), Prove, Nova 30/60, Multy, Move, 50 Tests
CAS Number:  
Price
171,00 €
Log in for availability
1 * 50 Tests
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*3 BOD nutrient salt mixture  1.00688 required
Specifications
Specifications
  • Chemical method:
    Winkler (modified)
  • Pk:
    50 Tests
  • Description:
    BOD cell test*³
  • For unit:
    Prove, Nova 30/60, Multy, Move
  • Range:
    0,5 - 3000 mg/l
  • Cat. No.:
    1.00687.0001
  • M-Clarity quality level:
    MQ100
Specifications
Accessories
Product Information
  • Catalog #
    Type
    Availability
    Price
  • Oxygen reaction tube for 1.14662.0001 (MTC)
    Oxygen reaction tube for 1.14662.0001 (MTC)
    1.14663.0001
    MTC
    Log in for availability
    1 * 1 items - 97,40 €