Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,3-Lutidine ≥99%

Glass bottle for solids, 500 g
Catalog # L03697.36
Toxic
Flammable
Irritant
2,3-Lutidine ≥99%
Catalog # L03697.36
CAS Number:  583-61-9
2,3-Lutidine ≥99%
2,3-Lutidine ≥99%

Glass bottle for solids, 500 g

CAS Number:  583-61-9
Price
259,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 583-61-9 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 50°(122°F) Refractive Index: 1.5080 Odour: Unpleasant Melting Point: -17° Boiling Point: 160-161° Beilstein Reference: 106418 UN No.: 2929 MDL No.: MFCD00009605

Synonyms:  2,3-Dimethylpyridine
  • CAS Number:583-61-9
  • Formula:C₇H₉N
  • MDL Number:MFCD00009605
  • Molecular Weight:107.16 g/mol
  • Boiling Pt:160…161 °C
  • Melting Pt:–17 °C
  • Density:0.948
  • Flash Pt:50 °C (122 °F)
  • Storage Temperature:Ambient
  • EINECS:209-514-1
  • UN:2929
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications