Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® Silicone Fluids

VWR® Silicone Fluids

Silicone fluid 20 cS, −40...+200 °C, 20 mm²/s, Clear, Plastic container, 20 kg
Catalog # 630057W
Supplier:   VWR Chemicals
VWR® Silicone Fluids
Catalog # 630057W
Supplier:  VWR Chemicals
CAS Number:  
VWR® Silicone Fluids
VWR® Silicone Fluids

Silicone fluid 20 cS, −40...+200 °C, 20 mm²/s, Clear, Plastic container, 20 kg

CAS Number:  
Price
7 850,00 €
Log in for availability
1 * 20 kg
Product Details & Documents

A range of silicone fluids recommended for use in baths. Low in toxicity, inert and long lasting due to their low volatility.

  • Non corrosive
  • High flash point
  • Long service life
  • Low evaporation

Fluids possess excellent heat transfer characteristics and cover a large temperature range from −40 to +200 °C.

1 cS = 1 mm²/s

Packaging: Packed in a plastic bottle/plastic container.

Caution: Harmful vapours may be given off at elevated temperatures. It is advisable to use this material in a fume cupboard at temperatures greater than 140 °C.

Specifications

Specifications

  • Pack type:
    Plastic container
  • Pk:
    20 kg
  • Colour:
    Clear
  • Description:
    Silicone fluid 20 cS
  • Temperature range:
    −40...+200 °C
  • Viscosity:
    20 mm²/s

Specifications