Order Entry
Export
Orders LinkContactUsLinkComponent
Burettes, removable stopcock

Burettes, removable stopcock

25 ml, 0,10 ml, 0,10 ± ml
Catalog # 612-0369
Supplier:   SciLabware
Burettes, removable stopcock
Catalog # 612-0369
Supplier:  SciLabware
Burettes, removable stopcock
Burettes, removable stopcock

25 ml, 0,10 ml, 0,10 ± ml

Price
80,80 €
Log in for availability
1 * 1 items
Product Details & Documents

Soda lime glass, class B, removable PTFE stopcock.

  • Flared top for safer filling
  • Blue enamel graduations

BS 846

Certifications: BS 846

Specifications
  • Capacity:
    25 ml
  • Seq:
    01
  • Division:
    0,10 ml
  • Tolerance:
    0,10 ± ml