Order Entry
Export
Orders LinkContactUsLinkComponent
Perylene ≥98%

Perylene ≥98%

Glass bottle for solids, 5 g
Perylene ≥98%
  L03047.06
 :  198-55-0
Perylene ≥98%
Perylene ≥98%

Glass bottle for solids, 5 g

 :  198-55-0
 
127,00 €
Log in for availability
1 * 5 g
 

Purity: 98+% CAS No.: 198-55-0 Molecular Formula: C20H12 Formula Weight: 252.32 Odour: Odourless Melting Point: 276-280° Beilstein Reference: 1911335 Merck Reference: 14,7184 MDL No.: MFCD00004142

  • CAS Number:198-55-0
  • Formula:C₂₀H₁₂
  • MDL Number:MFCD00004142
  • Molecular Weight:252.32 g/mol
  • Merck Index:14,7184
  • Boiling Pt:443 °C (1013 hPa)
  • Melting Pt:276…280 °C
  • Density:1.362 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-900-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications